Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdlim3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024376.1 | NP_001019547.1 | 6 - 7 | 795 | 89 | |
GenBank mRNA | BC095249.1 | - | 6 - 7 | 795 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |