Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flvcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025522.1 | NP_001020693.1 | 2 - 3 | 784 | 68 | |
XM_005160739.3 | XP_005160796.1 | 2 - 3 | 1211 | 68 | ||
XM_009294763.3 | XP_009293038.1 | 3 - 4 | 1092 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |