Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flvcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025522.1 | NP_001020693.1 | 8 - 9 | 1433 | 71 | |
XM_005160739.3 | XP_005160796.1 | 8 - 9 | 1860 | 71 | ||
XM_009294763.3 | XP_009293038.1 | 9 - 10 | 1741 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Danio rerio Probe spans exons |