Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rtn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029967.3 | NP_001025138.2 | 2 - 3 | 1203 | 64 | |
GenBank mRNA | AY555044.1 | - | 2 - 3 | 1106 | 64 | |
BC117568.1 | - | 2 - 3 | 1086 | 64 | ||
BC163936.1 | - | 2 - 3 | 1173 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |