Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rtn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029967.3 | NP_001025138.2 | 7 - 8 | 2566 | 71 | |
NM_001319289.1 | NP_001306218.1 | 5 - 6 | 552 | 71 | ||
NM_199652.2 | NP_955946.1 | 5 - 6 | 805 | 71 | ||
GenBank mRNA | AY555044.1 | - | 7 - 8 | 2469 | 71 | |
AY555046.1 | - | 7 - 8 | 1031 | 71 | ||
AY555049.1 | - | 5 - 6 | 644 | 71 | ||
AY555050.1 | - | 5 - 6 | 681 | 71 | ||
BC059623.1 | - | 5 - 6 | 659 | 71 | ||
BC071446.1 | - | 5 - 6 | 602 | 71 | ||
BC090486.1 | - | 5 - 6 | 1097 | 71 | ||
BC163936.1 | - | 7 - 8 | 2536 | 71 | ||
EH444584.1 | - | 5 - 6 | 755 | 71 | ||
EH487664.1 | - | 5 - 6 | 517 | 71 | ||
EH494741.1 | - | 5 - 6 | 578 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |