Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sept4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037379.1 | NP_001032456.1 | 9 - 10 | 1340 | 84 | |
GenBank mRNA | BC109428.1 | - | 9 - 10 | 1340 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |