Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfpm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039524.1 | NP_001034613.1 | 1 - 2 | 299 | 60 | |
XM_005159561.4 | XP_005159618.1 | 1 - 2 | 293 | 60 | ||
GenBank mRNA | AY515852.1 | - | 1 - 2 | 307 | 60 | |
BC163559.1 | - | 1 - 2 | 244 | 60 | ||
DQ015977.1 | - | 1 - 2 | 299 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |