Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zdhhc5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039640.1 | NP_001034729.1 | 4 - 5 | 1062 | 78 | |
XM_005160244.4 | XP_005160301.1 | 5 - 6 | 2546 | 78 | ||
XM_005160246.4 | XP_005160303.1 | 5 - 6 | 2546 | 78 | ||
XM_005160247.4 | XP_005160304.1 | 5 - 6 | 2546 | 78 | ||
XM_009293347.3 | XP_009291622.1 | 5 - 6 | 2546 | 78 | ||
GenBank mRNA | AY871210.1 | - | 4 - 5 | 1062 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |