Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zdhhc5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039640.1 | NP_001034729.1 | 9 - 10 | 1616 | 64 | |
GenBank mRNA | AY871210.1 | - | 9 - 10 | 1616 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |