Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nphs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040687.1 | NP_001035777.1 | 17 - 18 | 2292 | 81 | |
XM_017351014.2 | XP_017206503.1 | 19 - 20 | 2884 | 81 | ||
XM_021481465.1 | XP_021337140.1 | 18 - 19 | 2768 | 81 | ||
GenBank mRNA | DQ466904.1 | - | 17 - 18 | 2292 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |