Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nphs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040687.1 | NP_001035777.1 | 23 - 24 | 3118 | 94 | |
XM_017351014.2 | XP_017206503.1 | 25 - 26 | 94 | |||
XM_021481465.1 | XP_021337140.1 | 24 - 25 | 94 | |||
GenBank mRNA | DQ466904.1 | - | 23 - 24 | 3118 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |