Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plekhg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328211.1 | NP_001315140.1 | 9 - 10 | 1700 | 77 | |
XM_005169952.4 | XP_005170009.1 | 9 - 10 | 1897 | 77 | ||
XM_009293507.3 | XP_009291782.1 | 9 - 10 | 1894 | 77 | ||
GenBank mRNA | BC124825.1 | - | 3 - 4 | 306 | 77 | |
BC134844.1 | - | 4 - 5 | 447 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |