Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shdb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045077.2 | NP_001038542.1 | 5 - 6 | 990 | 97 | |
XM_005170476.4 | XP_005170533.1 | 6 - 7 | 1437 | 97 | ||
XM_009296270.3 | XP_009294545.1 | 7 - 8 | 1516 | 97 | ||
XM_021469342.1 | XP_021325017.1 | 7 - 8 | 1573 | 97 | ||
GenBank mRNA | AW154797.1 | - | 1 - 2 | 182 | 97 | |
BC097159.1 | - | 6 - 7 | 1337 | 97 | ||
BC124504.1 | - | 6 - 7 | 1411 | 97 | ||
BC129459.1 | - | 6 - 7 | 1429 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |