Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pglyrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045222.2 | NP_001038687.2 | 1 - 2 | 245 | 152 | |
GenBank mRNA | BC171349.1 | - | 1 - 2 | 245 | 152 | |
DQ447204.1 | - | 1 - 2 | 190 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |