Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pglyrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045222.2 | NP_001038687.2 | 3 - 4 | 1235 | 80 | |
GenBank mRNA | BC171349.1 | - | 3 - 4 | 1235 | 80 | |
DQ447204.1 | - | 3 - 4 | 1180 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |