Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fdx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076664.2 | NP_001070132.1 | 2 - 3 | 267 | 70 | |
GenBank mRNA | BC124384.1 | - | 2 - 3 | 248 | 70 | |
EH493611.1 | - | 2 - 3 | 314 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |