Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfcu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082899.1 | NP_001076368.1 | 2 - 3 | 533 | 76 | |
GenBank mRNA | BC163066.1 | - | 2 - 3 | 567 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |