Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pex11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103120.1 | NP_001096590.1 | 1 - 2 | 401 | 68 | |
GenBank mRNA | BC151942.1 | - | 1 - 2 | 401 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |