Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: senp6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021468556.1 | XP_021324231.1 | 22 - 23 | 3074 | 79 | |
XM_021468557.1 | XP_021324232.1 | 21 - 22 | 3010 | 79 | ||
XM_021468558.1 | XP_021324233.1 | 22 - 23 | 3061 | 79 | ||
XM_021468559.1 | XP_021324234.1 | 22 - 23 | 3071 | 79 | ||
XM_021468560.1 | XP_021324235.1 | 21 - 22 | 2980 | 79 | ||
XM_021468561.1 | XP_021324236.1 | 21 - 22 | 2967 | 79 | ||
GenBank mRNA | BC134226.1 | - | 6 - 7 | 538 | 79 | |
BC154420.1 | - | 9 - 10 | 1083 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |