Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: chico | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001312685.1 | NP_001299614.1 | 7 - 8 | 670 | 60 | |
NM_130911.2 | NP_570986.1 | 6 - 7 | 919 | 60 | ||
GenBank mRNA | AF085218.1 | - | 6 - 7 | 881 | 60 | |
BC097097.1 | - | 7 - 8 | 670 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |