Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sema4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131049.1 | NP_571124.1 | 4 - 5 | 803 | 72 | |
XM_005168429.4 | XP_005168486.1 | 4 - 5 | 792 | 72 | ||
XM_017359054.2 | XP_017214543.1 | 4 - 5 | 771 | 72 | ||
XM_021481386.1 | XP_021337061.1 | 3 - 4 | 733 | 72 | ||
GenBank mRNA | AF073289.1 | - | 4 - 5 | 803 | 72 | |
BC078280.1 | - | 3 - 4 | 408 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |