Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131191.1 | NP_571266.1 | 3 - 4 | 315 | 109 | |
GenBank mRNA | Y17256.1 | - | 315 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |