Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131207.1 | NP_571282.1 | 2 - 3 | 429 | 126 | |
GenBank mRNA | AF001299.1 | - | 2 - 3 | 429 | 126 | |
BC162280.1 | - | 2 - 3 | 422 | 126 | ||
BC162303.1 | - | 2 - 3 | 422 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |