Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131676.2 | NP_571751.1 | 5 - 6 | 599 | 89 | |
GenBank mRNA | AF155576.1 | - | 5 - 6 | 601 | 89 | |
BC076475.1 | - | 5 - 6 | 614 | 89 | ||
BC165038.1 | - | 5 - 6 | 561 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |