Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173223.2 | NP_775330.2 | 4 - 5 | 727 | 75 | |
GenBank mRNA | AF262048.1 | - | 4 - 5 | 524 | 75 | |
AY091914.1 | - | 4 - 5 | 464 | 75 | ||
BC081509.1 | - | 4 - 5 | 727 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |