Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thbs3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173225.1 | NP_775332.1 | 17 - 18 | 2171 | 90 | |
GenBank mRNA | AF462310.1 | - | 17 - 18 | 2171 | 90 | |
BC085524.1 | - | 17 - 18 | 2169 | 90 | ||
BC165677.1 | - | 17 - 18 | 2121 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |