Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: f7i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173228.1 | NP_775335.1 | 7 - 8 | 748 | 110 | |
GenBank mRNA | AF515269.1 | - | 7 - 8 | 748 | 110 | |
BC094994.1 | - | 7 - 8 | 726 | 110 | ||
BC115091.1 | - | 7 - 8 | 732 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |