Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199636.1 | NP_955930.1 | 5 - 6 | 508 | 123 | |
XM_021478186.1 | XP_021333861.1 | 5 - 6 | 709 | 123 | ||
GenBank mRNA | BC059454.1 | - | 5 - 6 | 508 | 123 | |
BC071510.1 | - | 5 - 6 | 525 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |