Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vapa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199918.2 | NP_956212.2 | 309 | 99 | ||
XM_021470247.1 | XP_021325922.1 | 1 - 2 | 341 | 99 | ||
GenBank mRNA | BC061951.1 | - | 282 | 99 | ||
BC065663.1 | - | 309 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |