Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trim25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200175.1 | NP_956469.1 | 1 - 2 | 647 | 104 | |
XM_017356449.2 | XP_017211938.1 | 1 - 2 | 668 | 104 | ||
GenBank mRNA | BC045450.1 | - | 1 - 2 | 647 | 104 | |
BC165638.1 | - | 1 - 2 | 630 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |