Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mitd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200201.2 | NP_956495.2 | 3 - 4 | 667 | 83 | |
GenBank mRNA | BC045955.1 | - | 3 - 4 | 406 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |