Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200223.1 | NP_956517.1 | 5 - 6 | 1007 | 124 | |
XM_021470458.1 | XP_021326133.1 | 5 - 6 | 1859 | 124 | ||
XM_021470459.1 | XP_021326134.1 | 5 - 6 | 1846 | 124 | ||
XM_021470460.1 | XP_021326135.1 | 5 - 6 | 1390 | 124 | ||
GenBank mRNA | BC046049.1 | - | 5 - 6 | 1007 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |