Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a32a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200256.1 | NP_956550.1 | 6 - 7 | 1055 | 93 | |
GenBank mRNA | BC048057.1 | - | 6 - 7 | 1055 | 93 | |
BC164127.1 | - | 6 - 7 | 883 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |