Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: znrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200870.1 | NP_957164.1 | 4 - 5 | 1191 | 134 | |
GenBank mRNA | BC063239.1 | - | 4 - 5 | 1191 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |