Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201091.1 | NP_957385.1 | 1 - 2 | 186 | 60 | |
GenBank mRNA | BC044433.1 | - | 1 - 2 | 186 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |