Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dynlt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205608.1 | NP_991171.1 | 2 - 3 | 154 | 67 | |
GenBank mRNA | AY398423.1 | - | 2 - 3 | 154 | 67 | |
BC081386.1 | - | 2 - 3 | 90 | 67 | ||
BC164919.1 | - | 2 - 3 | 105 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |