Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sypl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212669.1 | NP_997834.1 | 2 - 3 | 134 | 76 | |
XM_017357386.2 | XP_017212875.1 | 1 - 2 | 92 | 76 | ||
GenBank mRNA | AY394944.1 | - | 2 - 3 | 134 | 76 | |
BC092895.1 | - | 2 - 3 | 139 | 76 | ||
BC152273.1 | - | 2 - 3 | 149 | 76 | ||
BC154773.1 | - | 2 - 3 | 129 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |