Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: mknk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212723.1 | NP_997888.1 | 12 - 13 | 1318 | 89 | |
GenBank mRNA | BC045391.1 | - | 12 - 13 | 1318 | 89 | |
BC165349.1 | - | 11 - 12 | 1136 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |