Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212770.1 | NP_997935.1 | 7 - 8 | 1125 | 70 | |
GenBank mRNA | BC066689.1 | - | 7 - 8 | 1125 | 70 | |
BC154779.1 | - | 13 - 14 | 1527 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |