Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212988.1 | NP_998153.1 | 5 - 6 | 793 | 99 | |
XM_005159885.4 | XP_005159942.1 | 6 - 7 | 1034 | 99 | ||
GenBank mRNA | BC054680.1 | - | 5 - 6 | 793 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |