Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfand5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213039.1 | NP_998204.1 | 5 - 6 | 620 | 64 | |
XM_005171826.4 | XP_005171883.1 | 5 - 6 | 644 | 64 | ||
XM_021476056.1 | XP_021331731.1 | 5 - 6 | 808 | 64 | ||
XM_021476057.1 | XP_021331732.1 | 5 - 6 | 802 | 64 | ||
GenBank mRNA | BC059673.1 | - | 5 - 6 | 620 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |