Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps6kb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213076.1 | NP_998241.1 | 8 - 9 | 784 | 63 | |
GenBank mRNA | BC046888.1 | - | 8 - 9 | 784 | 63 | |
BC165362.1 | - | 8 - 9 | 739 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |