Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213172.1 | NP_998337.1 | 7 - 8 | 936 | 76 | |
GenBank mRNA | BC066368.1 | - | 7 - 8 | 936 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |