Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbwd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213253.1 | NP_998418.1 | 8 - 9 | 654 | 106 | |
GenBank mRNA | BC065429.1 | - | 8 - 9 | 654 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |