Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmed1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080640.1 | NP_001074109.1 | 5 | 948 | 118 | |
GenBank mRNA | BC129377.1 | - | 5 | 948 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |