Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077627.2 | NP_001071095.2 | 7 | 2479 | 148 | |
GenBank mRNA | EF014956.1 | - | 7 | 2446 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |