Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh2g28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024200.1 | NP_001019371.1 | 2 | 2450 | 134 | |
GenBank mRNA | AY583043.1 | - | 2 | 2450 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |