Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dio1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC076008.1 | - | 4 | 948 | 120 | |
BC155742.1 | - | 4 | 895 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |