Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hs3st3b1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076646.1 | NP_001070114.1 | 3 | 1254 | 116 | |
GenBank mRNA | BC124198.1 | - | 3 | 1254 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |