Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC068984.1 | - | 1450 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio |